missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XI Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568371
Additional Details : Weight : 0.01000kg
Description
Carbonic Anhydrase XI (CA11, CA XI) is a member of the carbonic anhydrases (CAs) family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Carbonic Anhydrase XI participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. Further, Carbonic Anhydrase XI show extensive diversity in tissue distribution and in their subcellular localization. CA XI is likely a secreted protein, however, radical changes at active site residues completely conserved in CA isozymes with catalytic activity, make it unlikely that it has carbonic anhydrase activity. It shares properties in common with two other acatalytic CA isoforms, CA VIII and CA X. CA XI is most abundantly expressed in brain, and may play a general role in the central nervous system.Specifications
Carbonic Anhydrase XI | |
Polyclonal | |
Unconjugated | |
CA11 | |
CA11; Car11; carbonic anhydrase 11; carbonic anhydrase XI; carbonic anhydrase-related protein 11; Carbonic anhydrase-related protein 2; carbonic anhydrase-related protein XI; carbonic anhydrase-related XI protein; CA-RP II; CARP XI; CA-RP XI; Carp2; CARP-2; CARPX1; CAR-XI; CA-XI; UNQ211/PRO237 | |
Rabbit | |
affinity chromatography | |
RUO | |
770 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O75493 | |
CA11 | |
Synthetic protein corresponding to residues near the C terminal of human Carbonic Anhydrase XI. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |