Learn More
Carbonic Anhydrase XII Monoclonal Antibody (CL0280), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA524597
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TASNKSEGLA VLAVLIEMGS FNPSYDKIFS HLQHVKYKGQ EAFVPGFNIE ELLPERTAEY YRYRGSLTTP PCNPTVLWTV FRNPVQISQE QLLALETALY CTHMDDPSPR EMINNFRQVQ Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%. Binds to an epitope located within the peptide sequence HLQHVKYKGQEAFVP as determined by overlapping synthetic peptides.
Carbonic Anhydrase XII is a protein coding gene.Specifications
Carbonic Anhydrase XII | |
Monoclonal | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43570 | |
CA12 | |
Recombinant protein corresponding to Human Carbonic Anhydrase XII | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
CL0280 | |
Unconjugated | |
CA12 | |
2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3 | |
Mouse | |
Protein A | |
RUO | |
771 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only