Learn More
Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min, Thermo Scientific Chemicals
Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation
£41.20 - £1209.00
Chemical Identifiers
CAS | 17185-29-4 |
---|---|
Molecular Formula | C55H46OP3Rh |
Molecular Weight (g/mol) | 918.80 |
MDL Number | MFCD00151644 |
InChI Key | AACIZACVKFEETJ-UHFFFAOYSA-N |
Synonym | tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i |
PubChem CID | 132451836 |
IUPAC Name | carbon monoxide;rhodium;triphenylphosphane |
SMILES | [RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316717
|
Thermo Scientific Alfa Aesar
010016.77 |
0.1 g |
£41.20
0.10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306717
|
Thermo Scientific Alfa Aesar
010016.03 |
1 g |
£254.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396707
|
Thermo Scientific Alfa Aesar
010016.06 |
5 g |
£1209.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17185-29-4 | |
918.80 | |
AACIZACVKFEETJ-UHFFFAOYSA-N | |
132451836 | |
[RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C55H46OP3Rh | |
MFCD00151644 | |
tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i | |
carbon monoxide;rhodium;triphenylphosphane |
Specifications
17185-29-4 | |
Yellow | |
Odorless | |
RhH(CO)[P(C6H5)3]3 | |
Moisture sensitive | |
tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i | |
AACIZACVKFEETJ-UHFFFAOYSA-N | |
carbon monoxide;rhodium;triphenylphosphane | |
132451836 | |
Rh 10.0% min. | |
Carbonylhydridotris(triphenylphosphine)rhodium(I) |
138°C (decomposition) | |
1.33 g/mL | |
C55H46OP3Rh | |
MFCD00151644 | |
14,9758 | |
Soluble in hydrocarbons (e.g. benzene and toluene) with dissociation of phosphine ligands | |
[RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
918.80 | |
918.8 | |
Powder |
Safety and Handling
P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
H312+H332
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 241-230-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only