missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Carboxymethylecellulose, sodium salt, average M.W. 90000 (DS=0.7)
£24.62 - £439.19
Chemical Identifiers
CAS | 9004-32-4 |
---|---|
Molecular Formula | (C12 H14 O9 R6)n |
Molecular Weight (g/mol) | 263.20 |
MDL Number | MFCD00081472 |
InChI Key | DPXJVFZANSGRMM-UHFFFAOYNA-N |
Synonym | carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn |
PubChem CID | 23706213 |
SMILES | [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10717222
|
Thermo Scientific Chemicals
332601000 |
100 g | Glass bottle |
£24.62
100g |
In Stock
Log in to see stock. |
||||
10429681
|
Thermo Scientific Chemicals
332600010 |
1 kg | Plastic bottle |
£104.52
1kg |
In Stock
Log in to see stock. |
||||
11986981
|
Thermo Scientific Chemicals
332600050 |
5 kg | Plastic drum |
£439.19
5kg |
Estimated Shipment: 05-12-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9004-32-4 | |
263.20 | |
DPXJVFZANSGRMM-UHFFFAOYNA-N | |
23706213 |
(C12 H14 O9 R6)n | |
MFCD00081472 | |
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn | |
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
Specifications
White to Yellow | |
9004-32-4 | |
>300.0°C | |
MFCD00081472 | |
DPXJVFZANSGRMM-UHFFFAOYNA-N | |
sodium;2,3,4,5,6-pentahydroxyhexanal;acetate | |
23706213 | |
Authentic | |
50-100 mPa.s 2% sol. |
6.5 to 8 (1% soln.) | |
Granular Powder | |
(C12 H14 O9 R6)n | |
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn | |
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O | |
263.20 | |
≥99.5% | |
10% max. (as packed) (3 to 5 g, 105°C, 2 hrs) | |
Carboxymethylecellulose, sodium salt, Average M.W. 90000 (DS=0.7) |
RUO – Research Use Only