missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
£140.00 - £352.00
Specifications
Antigen | Carboxypeptidase A4 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17733564
|
Invitrogen
PA5122165 |
20 μL |
£140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17723564
|
Invitrogen
PA5122164 |
100 μL |
£352.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carboxypeptidase A4 is a member of the carboxypeptidase A/B subfamily. Carboxypeptidases are zinc-containing exopeptidases that catalyze the release of carboxy-terminal amino acids, and are synthesized as zymogens that are activated by proteolytic cleavage. This protein could be involved in the histone hyperacetylation pathway. It is imprinted and may be a strong candidate protein for prostate cancer aggressiveness.Specifications
Carboxypeptidase A4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UI42 | |
51200 | |
Lys55-Tyr421 (Accession Q9UI42), with N-terminal His Tag | |
Primary | |
-20°C | |
CPA4 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
1110019K20Rik; AV009555; Carboxypeptidase A3; carboxypeptidase A4; CPA3; Cpa4; RGD1563619; UNQ694/PRO1339 | |
CPA4 | |
IgG | |
Antigen affinity chromatography | |
Antibody |