Learn More
Carboxypeptidase B1 Recombinant Rabbit Monoclonal Antibody (329), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA529183
264.60 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Carboxypeptidase B1/CPB1.
Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
Specifications
Carboxypeptidase B1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P15086 | |
CPB1 | |
Recombinant Human Carboxypeptidase B1/CPB1 protein (Met1-Tyr417) | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
329 | |
Unconjugated | |
CPB1 | |
0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870; carboxypeptidase B; carboxypeptidase B1; carboxypeptidase B1 (tissue); Cpb; Cpb1; Pancreas-specific protein; pancreatic carboxypeptidase B; pancreatic carboxypeptidase B1; PASP; PCPB; procarboxypeptidase B; protaminase; tissue carboxypeptidase B | |
Rabbit | |
Protein A | |
RUO | |
1360 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.