Learn More
Wesbart™ Cardboard Cryo Boxes
Wesbart™ Cardboard Cryo Boxes are supplied with lids and have a water repellent coating for extra durability
Brand: Wesbart™ A3
This item is not returnable.
View return policy
Description
- For storing items loose or cryovials with dividers
- Sturdy fibreboard construction
- Water repellent coating for durability
- Supplied with lids
- Dividers sold separately
- Recyclable
Specifications
White | |
Cryo Box | |
Loose Items or Cryovials (With Dividers) | |
A3 |
Cardboard | |
Cryogenic Storage | |
Lid | |
133 x 133 x 100 mm |
Product Content Correction
Product Title
Wesbart™ Cardboard Cryo Boxes > 133 x 133 x 100 mm
Spot an opportunity for improvement?Share a Content Correction