missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Cardboard Rack for Upright Freezers, 50 mm Box Height

Due to the design and choice of material, these unique racks are extremely solid and long lasting.

Brand:  Tenak™ TE26134

 View more versions of this product

Product Code. 15280994

  • £43.50 / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gives the end user a second to none opportunity for reducing costs
  • Ideal for long term storage or for applications where price is of high priority
Specifications

Specifications

50 mm (Box)
Upright Freezers
5 x 4
562 x 139 x 285 mm
Boxes (20)
Cardboard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers