missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cargille Immersion Oil
Greatly increases light-gathering power with high-power objectives
Description
Greatly increases light-gathering power with high-power objectives
- Formulated to give refractive index of 1.5150 ± 0.0002 for D line at 23°C (73°F)
- Nondrying, so index will not change when exposed to air
- Contains no polychlorinated biphenyl (PCB) compounds
- Conforms to ISO 8036/1 specifications
- Standard for all oil-immersion microscopy
- Type A oil has low viscosity and low fluorescence
- Type B is a high-viscosity oil used in biomedical microscopy
- Type NVH is a high-viscosity oil that meets requirements for horizontal, inverted and inclined instruments and projection equipment
- Type FF oil is virtually nonfluorescent
- Type DF is a very low-fluorescence oil for fluorescence microscopy with very high resolution
Specifications
ISO 8036/1 | |
1.5150 ±0.0002 |
Safety and Handling
Product Identifier
Signal Word
- Cargille Immersion Oil Type B
Signal Word
- Warning
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cargille Immersion Oil