missing translation for 'onlineSavingsMsg'
Learn More

Carminic acid, 70-90%, Thermo Scientific Chemicals

Product Code. 10215781
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1260-17-9
Molecular Formula C22H20O13
Molecular Weight (g/mol) 492.39
MDL Number MFCD00167028
InChI Key DGQLVPJVXFOQEV-NGOCYOHBSA-N
Synonym C.I. 75470, Natural Red 4
PubChem CID 10255083
ChEBI CHEBI:78310
IUPAC Name 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid
SMILES CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C([C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=C(O)C(O)=C1C2=O

Specifications

Assay Percent Range 70-90%
Beilstein 10, II, 776
Merck Index 15, 1843
Infrared Spectrum Authentic
Formula Weight 492.39
Percent Purity ≥70%
Packaging Glass bottle
Color Red
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Carminic acid, 70-90%
Show More Show Less
compliance-icons
Product Identifier
  • Carminic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'