Learn More
Thermo Scientific™ Carrying Kit Set for Eutech™ Cond 6+ and TDS 6+ meters
Eutech™ Carrying Kit Cases are thoughtfully designed to hold your Eutech™ meter and electrodes.
Brand: Thermo Scientific™ 01X266902
Code : 04
Additional Details : Weight : 0.10000kg
Description
Eutech™ Carrying Kit Cases are thoughtfully designed to hold Eutech™ meters and electrodes.The Economy Carrying Kit Set is a plastic carrying case for the Eutech™ Cond 6+ and TDS 6+ meters and probes, calibration solutions, batteries, and manuals.
The Carrying Kit set is comprised of:
- 1413 μS/CM KCL Calibration Solution
Specifications
Carrying Kit Set for Eutech™ Cond 6+ and TDS 6+ meters | |
Accessory | |
1413 μS/CM KCL Calibration Solution, 12.88 mS/cm KCl Calibration Solution, 3000 ppm 442 TDS standard solutions, deionised rinse water (60 ml) | |
Eutech™ Cond 6+ and TDS 6+ meters |