Learn More
Bel-Art™ Scienceware™ Carton Biohazard Incineration
Perfect for containing biohazardous waste slated for incineration. Bel Art™ Carton Biohazard Incineration cartons can be incinerated by placing the entire unit in the incinerator.
Brand: Bel-Art™ Scienceware™ F13205-0002
73.75 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Code : 02
Additional Details : Weight : 1.91700kg
Description
- Tabs along the top edge keep a clear 0.038mm (1.5 mil) thick polypropylene biohazard bag from slipping out of place
- Snug-fitting lid has a flap for easily inserting waste material
- Printed with large, bright, biohazard graphics and instructions in four languages (English, Spanish, French, German)
- Packaged flat to save space and shipping costs; carton and inner bag can be assembled in seconds
- Available in sizes to suit benchtop or floor use
- Optional, reusable, sanitary cover (F13204-0000) for floor model sold separately
Specifications
250 mm | |
0.04 mm | |
Carton Biohazard Incineration |