Learn More
BUCHI Cartridges
Brand:  BUCHI 044883
Product Code. 12896893
  • POA / Pack of 50
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Specifications
150 mm
Empty Cartridge
Cartridger C-670 for Packing
Polypropylene
50 frits for packing of 25 cartridges
up to 10 bar
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BUCHI Cartridges > Dimensions: 40 dia. x 150mmL