missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear CAS Safety Shoes
Washable microfiber uppers are treated to repel all kinds of dirt, germs and bacteria. A highly breathable, comfortable shoe for all day wear. Energy returning soles prevent fatigue and keep the wearer fresh, alert, foot sure and steady even in the event of a spillage.
£122.96 - £124.02
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | White |
Design | CAS |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15958661
|
Emma Safety Footwear
MM52054035 |
35 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15968661
|
Emma Safety Footwear
MM52054036 |
36 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15978661
|
Emma Safety Footwear
MM52054037 |
37 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15988661
|
Emma Safety Footwear
MM52054038 |
38 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15998661
|
Emma Safety Footwear
MM52054039 |
39 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15908671
|
Emma Safety Footwear
MM52054040 |
40 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15918671
|
Emma Safety Footwear
MM52054041 |
41 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15928671
|
Emma Safety Footwear
MM52054042 |
42 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15938671
|
Emma Safety Footwear
MM52054043 |
43 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15948671
|
Emma Safety Footwear
MM52054044 |
44 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15958671
|
Emma Safety Footwear
MM52054045 |
45 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15968671
|
Emma Safety Footwear
MM52054046 |
46 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15978671
|
Emma Safety Footwear
MM52054047 |
47 |
£124.02
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
15988671
|
Emma Safety Footwear
MM52054048 |
48 |
£122.96
1 pair |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||||
Description
- S2-Safety shoe for wet working conditions
- GripForce™ Easy Twist Outsole
- Microfiber construction
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011 | |
White | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Laces | |
CAS | |
Microfiber |