missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-3 Antibody - (active/cleaved), Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 48 publications
Brand: Novus Biologicals NB100-56113
This item is not returnable.
View return policy
Description
Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular).
Specifications
| Caspase-3 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Flow Cytometry reported in scientific literature (PMID 28287161), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29963272), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen reported in scientific literature (PMID 29864441), Immunoblotting reported in scientific literature (PMID 27912032), Flow (Intracellular) reported in scientific literature (PMID 24804954) | |
| Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| Rabbit | |
| 31.7 kDa | |
| 0.05 mL | |
| Apoptosis, Cancer, Caspases, Phospho Specific | |
| 836 | |
| Human, Mouse, Rat, Gerbil | |
| Antisera |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| Unconjugated | |
| Neat whole antisera with 0.05% Sodium Azide | |
| CASP3 | |
| This Caspase-3 Antibody - (active/cleaved) was developed against catalytically active human caspase-3 protein. | |
| Unpurified | |
| RUO | |
| Primary | |
| Preferentially detects active caspase-3 (large subunit: ∼14-21 kDa, and small subunit: ∼10 kDa). However, it may also detect pro-caspase-3 (∼32 kDa) in some cell or tissue systems. Nuclear immunostaining of caspase-3 is considered be an indication of active/cleaved caspase-3. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction