Learn More
Epredia™ Cassette 1 Slotted Tissue Cassettes
Hold tissue specimens during processing and embedding with Epredia™ Cassette 1 slotted tissue cassettes.
Brand:  Epredia™ 1000961
 View more versions of this product
Product Code. 15327260
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Acetal polymer construction
  • 30° writing face
  • Small slot-style pores
Ordering Alerts:
  • Supplied in box of 500 cassettes with attached breakaway lids (not available in Tube Packs)
Specifications
Specifications
Slotted Pore
Blue
Tissue Cassette
30°
Acetal Polymer