Learn More
Cassette Clamp, right
Cassette clamps are replacement parts for the Mini Gel Tank
Brand: Invitrogen
Code : NEW
Additional Details : Weight : 0.40000kg
Description
Cassette clamps are replacement parts for the Mini Gel Tank. They lock and seal the gel cassettes within the Gel Runner Tank. The orientation is determined when facing the front of the gel tank.About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Universal | |
Mini | |
Room Temperature |
Novex™ Mini Gels | |
Cassette Clamp |
For Research Use Only. Not for use in diagnostic procedures.