Learn More
Epredia™ Cassette II Slotted Tissue Cassettes
For routine tissue samples, these cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Brand:  Epredia™ B851739LA
 View more versions of this product
Product Code. 16384136
  • £210.00 / Pack of 1000
Explore more special offers
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
Slots
1000/Cs.
Lavender
Yes
Separate Lid, Front Hinge
Slotted Cassette