Learn More
Epredia™ Cassette II Slotted Tissue Cassettes
For routine tissue samples, these cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Brand: Epredia™ B851739LG
150.75 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Slots | |
1000/Cs. | |
Light Green |
Yes | |
Separate Lid, Front Hinge | |
Slotted Cassette |