Learn More
Epredia™ Cassette II Slotted Tissue Cassettes in Tube Packs
Slotted Cassettes are ideal for routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a front edge lid hinge, and loose-packed lids that are available in 12 color varieties.
Brand:  Epredia™ B851729TN
 View more versions of this product
Product Code. 16304066
  • £118.00 / Pack of 750
Explore more special offers
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
Slots
750/Cs.
Yes
For Use With PrintMate™ Printers
Yes
Separate Lid, Front Hinge
Tan
Cassette II Biopsy Cassette