Learn More
R&L Enterprises™ Tripod Shape Retort Stand Base
Excellent option for supporting retort stands in the laboratory. R&L Enterprises™ Tripod Shape Retort Stand Base are made from cast iron.
Brand:  R&L Enterprises™ 3025/3
 View more versions of this product
Product Code. 12338139
  • £48.80 / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Painted blue
  • With rubber feet
  • Central rod hole is tapped
  • Without rod
Specifications
Specifications
Cast Iron
With Rubber Feet
Blue
Rod Centre: 135mm
Tripod Spread Base
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products