missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Castor oil, specified according to the requirements of Ph.Eur.
£21.50 - £73.10
Chemical Identifiers
CAS | 8001-79-4 |
---|---|
Molecular Formula | C57H104O9 |
Molecular Weight (g/mol) | 933.45 |
MDL Number | MFCD00130746 |
InChI Key | ZEMPKEQAKRGZGQ-AAKVHIHISA-N |
Synonym | castor oil, ricinus oil, olio di ricino, venelex, xenaderm, optase, trypsin complex, unii-d5340y2i9g, castor oil usp:jan, 1-o,2-o,3-o-tris z-12-hydroxy-9-octadecenoyl glycerol |
PubChem CID | 14030006 |
IUPAC Name | 2,3-bis[[(Z)-12-hydroxyoctadec-9-enoyl]oxy]propyl (Z)-12-hydroxyoctadec-9-enoate |
SMILES | CCCCCCC(CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(CCCCCC)O)OC(=O)CCCCCCCC=CCC(CCCCCC)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10566051
|
Thermo Scientific Chemicals
348855000 |
500 mL | Glass bottle |
£21.50
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974041
|
Thermo Scientific Chemicals
348850025 |
2.5 L | Glass bottle |
£73.10
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
8001-79-4 | |
933.45 | |
ZEMPKEQAKRGZGQ-AAKVHIHISA-N | |
14030006 | |
CCCCCCC(CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(CCCCCC)O)OC(=O)CCCCCCCC=CCC(CCCCCC)O)O |
C57H104O9 | |
MFCD00130746 | |
castor oil, ricinus oil, olio di ricino, venelex, xenaderm, optase, trypsin complex, unii-d5340y2i9g, castor oil usp:jan, 1-o,2-o,3-o-tris z-12-hydroxy-9-octadecenoyl glycerol | |
2,3-bis[[(Z)-12-hydroxyoctadec-9-enoyl]oxy]propyl (Z)-12-hydroxyoctadec-9-enoate |
Specifications
2.0mg KOH/g max. | |
1.5 max. (at the absorption maximum at 269 +/- 1nm) | |
Colorless or Yellow | |
229.0°C | |
Authentic | |
1.4770 to 1.4810 | |
14,1898 | |
+ 3.50 | |
Solubility in water: .. Other solubilities: miscible with alcohol, benzene, chloroform,, carbondisulfide, ether and glacial acetic acid | |
CCCCCCC(CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(CCCCCC)O)OC(=O)CCCCCCCC=CCC(CCCCCC)O)O | |
933.45 | |
0,6-0,8 Pas (25°C) | |
of fatty acids (GC): Passes Test (per Eur. Pharm.) | |
Castor oil |
8001-79-4 | |
-10.0°C | |
0.9570g/mL | |
229°C | |
C57H104O9 | |
MFCD00130746 | |
0.957 | |
castor oil, ricinus oil, olio di ricino, venelex, xenaderm, optase, trypsin complex, unii-d5340y2i9g, castor oil usp:jan, 1-o,2-o,3-o-tris z-12-hydroxy-9-octadecenoyl glycerol | |
ZEMPKEQAKRGZGQ-AAKVHIHISA-N | |
2,3-bis[[(Z)-12-hydroxyoctadec-9-enoyl]oxy]propyl (Z)-12-hydroxyoctadec-9-enoate | |
14030006 | |
Eu. Ph. | |
Viscous Hygroscopic Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 232-293-8
missing translation for 'rtecsNumber' : FI4100000
missing translation for 'tsca' : TSCA
RUO – Research Use Only