missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Castors with Plinth

65 mm castors with plinth

Brand:  asecos™ 28082

 View more versions of this product

Product Code. 11928837

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Alert:

Use only in combination with cabinet

This item is not returnable. View return policy

Description

Description

  • Sheet steel powder-coated
  • For model UB90
  • For under bench cabinets
  • Height = 65 mm
Specifications

Specifications

UB90 with a width of 1100 mm
Powder-coated plain
Steel
Model UB90 under bench cabinets
3 kg
Castors with plinth
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers