Learn More
Fisherbrand™ Catalyst PharMed BPT Two-Stop Microbore Tubing Sets
Preconfigured ready-to-use links and assemblies.
Brand:  Fisherbrand™ 97200-23
 View more versions of this product
Product Code. 15511385
  • £186.00 / Pack of 12
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For small cartridges
  • Time-saving assembly
  • Four formulations for range of chemical compatibility
  • Use with Catalyst extension coils/tubing
Specifications
Specifications
Two-stop microbore tubing set
0.25 mm