Learn More
Fisherbrand™ Catalyst Platinum-Cured Silicone Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Brand:  Fisherbrand™ 97100-13
 View more versions of this product
Product Code. 15501365
  • £230.00 / Pack of 6
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Preconfigured ready-to-use links and assemblies.
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head
Specifications
Specifications
1.6 mm
Platinum-cured Silicone
1.6 mm
Catalyst extension coils and tubing