Learn More
CatecholsulfonpHthalein, Pure, Metal Indicator, Fisher Chemical
Brand: Fisher Chemical C/3360/44
Code : 21
Additional Details : CAS Number : 115-41-3 Weight : 0.00500kg
Specifications
115-41-3 | |
5868 | |
Glass Jar | |
5 g |
C19H14O7S | |
386.36g/mol | |
Green | |
Powder |