Learn More
Astell Scientific™ Category 3 (BSL-3) Compliance Option
Required when dealing with high risk sterilization requirements. Astell Scientific™ Category 3 (BSL-3) Compliance Option uses a bacterial retentive filter fitted onto the exhaust to ensure that nothing leaves the chamber without being sterilized.
Brand: Astell Scientific™ AVQ005
This item is not returnable.
View return policy
Description
- Required for high risk sterilization processes.
- Bacterial retentive filter.
- Prevents anything from leaving the autoclave chamber without being sterilized.
Specifications
Category 3 (BSL-3) | |
High-Risk Sterilization Processes | |
33, 43, 63 L Compact Autoclaves |