missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin H Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547041
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) Cathepsin H is observed and less than 2% cross-reactivity with recombinant mouse (rm) Cathepsin B, rmCathepsin C, rmCathepsin Z, rmCathepsin L, and rhCathepsin O is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Cathepsin H Polyclonal antibody specifically detects Cathepsin H in Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
Cathepsin H | |
Polyclonal | |
Unconjugated | |
CTSH | |
ACC4; ACC-4; ACC5; ACC-5; AL022844; aleurain; Cat H; CATH; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; CPSB; Ctsh; N-benzoylarginine-beta-naphthylamide hydrolase; pro-cathepsin H | |
Goat | |
Antigen affinity chromatography | |
RUO | |
13036 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P49935 | |
CTSH | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin H Glu22-Val333 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |