missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CATSPER1 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577359
Additional Details : Weight : 0.01000kg
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Voltage-gated calcium channel that plays a central role in calcium-dependent physiological responses essential for successful fertilization, such as sperm hyperactivation, acrosome reaction and chemotaxis towards the oocyte. Activated by extracellular progesterone and prostaglandins following the sequence: progesterone > PGF1-alpha = PGE1 > PGA1 > PGE2 >> PGD2. The primary effect of progesterone activation is to shift voltage dependence towards more physiological, negative membrane potentials; it is not mediated by metabotropic receptors and second messengers. Sperm capacitation enhances the effect of progesterone by providing additional negative shift. Also activated by the elevation of intracellular pH.Specifications
CATSPER1 (extracellular) | |
Polyclonal | |
Unconjugated | |
Catsper1 | |
CatSper1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
225865, 689349 | |
-20°C | |
Lyophilized |
Western Blot | |
0.6 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q91ZR5 | |
Catsper1 | |
Peptide (C)RALFQDSDPKRFQN, corresponding to amino acid residues 508-521 of mouse CatSper1 | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |