missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14787-1-AP-20UL
This item is not returnable.
View return policy
Description
Strongly expressed in human liver and pancreas, with weaker expression in heart and brain, the cytoplasmic protein cystathionine b-synthase (CBS) operates in the first step of homocysteine transulfuration. CBS, which belongs to the cysteine synthase/cystathionine b-synthase family of proteins, catalyzes the formation of cystathionine from the thrombogenic amino acid homocysteine using pyridoxal phosphate cofactor. Allosteric activation by adenosyl-methionine regulates CBS activity. Deficiencies in CBS are assoc-iated with homocystinuria, a recessively inherited error in sulfur amino acid metabolism that affects many organs and tissues. Symptoms of homocytinuria include arteriosclerosis, thrombosis, dislocated optic lenses, mental retard-ation and skeletal abnormalities.Specifications
CBS | |
Polyclonal | |
Unconjugated | |
CBS | |
Beta thionase, CBS, cystathionine beta synthase, HIP4, Serine sulfhydrase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12411, 24250, 875 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32232, P35520, Q91WT9 | |
CBS | |
CBS Fusion Protein Ag6437 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |