missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCAR2 Mouse anti-Human, Porcine, Clone: 2D10F8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | CCAR2 |
---|---|
Clone | 2D10F8 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888425
|
Proteintech
66497-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878425
|
Proteintech
66497-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DBC1 is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5′ CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.Specifications
CCAR2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CCAR2, cell cycle and apoptosis regulator 2, DBC 1, DBC.1, DBC1, KIAA1967, p30 DBC, Protein KIAA1967 | |
CCAR2 | |
IgG2a | |
Protein A | |
Antibody |
2D10F8 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Porcine, Human | |
Q8N163 | |
100155718, 57805 | |
CCAR2 Fusion Protein Ag18374 | |
Primary | |
-20°C | |
CCAR2 |