missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CCDA Selective Medium
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
CCDA Selective Medium
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
Description
Isolate Campylobacter spp. using blood-free Thermo Scientific™ CCDA Selective Medium, which conforms to ISO 10272-1 and 10272-2.
Modified CCDA Medium is based on the original formulation described by Bolton which was developed to replace blood with charcoal, ferrous sulphate and sodium pyruvate. Improved selectivity is achieved using cefoperazone as the selective agent. Amphotericin B has been added to the formula to suppress the growth of yeast and fungal contaminants that may occur at 37°C.
Campylobacter infections are among the most common bacterial infections, producing both gastrointestinal and systemic illnesses. Campylobacter jejuni is the most commonly reported bacterial cause of foodborne infection in the United States. Campylobacter are small, non-sporeforming, helical, curved gram-negative rods that require a microaerophilic environment and cannot tolerate drying.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Campylobacter infections are among the most common bacterial infections, producing both gastrointestinal and systemic illnesses. Campylobacter jejuni is the most commonly reported bacterial cause of foodborne infection in the United States. Campylobacter are small, non-sporeforming, helical, curved gram-negative rods that require a microaerophilic environment and cannot tolerate drying.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Certifications/Compliance | According ISO 10271-a and ISO 10272-2 |
| Color | Black, opaque |
| Content And Storage | 2°C to 10°C |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Media Type | CCDA Selective Medium |
| Product Type | Agar |
| Fill Weight | 19.5g ± 1.0g |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction