Learn More
CCDC28A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14807-1-AP-20UL
 View more versions of this product
Product Code. 16883634
  • £135.00 / 20µL
Explore more special offers
Description
Description
This gene encodes a coiled-coil domain containing protein. Although the specific function of this gene has not yet been determined, this gene is a known translocation partner of nucleoporin 98 in acute leukemias. The resulting fusion gene produces a nucleoporin 98-coiled-coil domain-containing protein 28A chimeric protein which may be involved in promoting myeloproliferative neoplasms.
Specifications
Specifications
CCDC28A
Polyclonal
Unconjugated
CCDC28A
C6orf80, CCDC28A, CCRL1AP
Rabbit
Antigen Affinity Chromatography
RUO
215814, 25901, 361454
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IWP9
CCDC28A
CCDC28A Fusion Protein Ag6529
20 μL
Primary
Rat, Human, Mouse