missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC28A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | CCDC28A |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883634
|
Proteintech
14807-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873634
|
Proteintech
14807-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a coiled-coil domain containing protein. Although the specific function of this gene has not yet been determined, this gene is a known translocation partner of nucleoporin 98 in acute leukemias. The resulting fusion gene produces a nucleoporin 98-coiled-coil domain-containing protein 28A chimeric protein which may be involved in promoting myeloproliferative neoplasms.Specifications
CCDC28A | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q8IWP9 | |
215814, 25901, 361454 | |
CCDC28A Fusion Protein Ag6529 | |
Primary | |
-20°C | |
CCDC28A |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C6orf80, CCDC28A, CCRL1AP | |
CCDC28A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |