Learn More
CCDC93 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20861-1-AP-150UL
 View more versions of this product
Product Code. 16800445
  • £349.00 / 150µL
Explore more special offers
Description
Description
CCDC93 is involved in copper-dependent ATP7A trafficking between the trans-Golgi network and vesicles in the cell periphery; the function is proposed to depend on its association within the CCC complex and cooperation with the WASH complex on early endosomes and is dependent on its interaction with WASHC2C.
Specifications
Specifications
CCDC93
Polyclonal
Unconjugated
CCDC93
CCDC93
Rabbit
Antigen Affinity Chromatography
RUO
304743, 54520, 70829
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q567U6, Q5BJT7, Q7TQK5
CCDC93
CCDC93 Fusion Protein Ag14855
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
CCDC93 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated