Learn More
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J303, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Brand: Paragon Scientific CL09
Additional Details : CAS Number : 68037-01-4 Weight : 0.90000kg
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Manufactured in the United Kingdom
- 2 year shelf life
Specifications
Glass Amber Bottle | |
Yes | |
Liquid |
Safety and Handling
- CCS Viscosity Standard, CL090 (CL09)
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : Dec-1-ene, dimers, hydrogenated
Nominal values quoted. Certified Values will vary from batch to batch.