missing translation for 'onlineSavingsMsg'
Learn More

CD 1530, Tocris Bioscience™

Product Code. 18064302
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective RARγ agonist

Potent and selective RARγ receptor agonist (Ki values are 150, 1500 and 2750 nM for RARγ, RARβ and RARα receptors respectively). Activates transcriptional activity (AC50 = 1.8 nM).

Chemical Identifiers

CAS 107430-66-0
Molecular Formula C27H26O3
Molecular Weight (g/mol) 398.502
InChI Key VCQGNUWOMLYNNG-UHFFFAOYSA-N
Synonym cd hplc
PubChem CID 9952709
IUPAC Name 4-[7-(1-adamantyl)-6-hydroxynaphthalen-2-yl]benzoic acid
SMILES C1C2CC3CC1CC(C2)(C3)C4=C(C=C5C=CC(=CC5=C4)C6=CC=C(C=C6)C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 398.49
Percent Purity >98%
Chemical Name or Material CD 1530
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.