missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD 293 AGT™ Medium

Product Code. 11550436
Click to view available options
Quantity:
1 L
10 kg
10 L
100 L
Format:
Ropak Bucket
Securitainer Capped Vial
Unit Size:
1 L
10 L
100 L
10kg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11550436

Brand: Gibco™ 12529012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format.

CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format. CD 293 AGT™ is optimized for the growth of suspension cultures of 293 cells.

AGT = Advanced Granulation Technology

TRUSTED_SUSTAINABILITY

Specifications

Cell Line 293 (HEK)
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Powder (AGT)
Product Type CD 293 Medium
Serum Level Serum-free
With Additives Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine
Quantity 10 L
Shipping Condition Wet Ice
Endotoxin Level Low (≤10 EU/mL)
Format Securitainer Capped Vial
Included Antibiotics No antibiotics
Reconstituted Volume 10 L
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light.
Culture Type Suspension Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.