missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CD CHO AGT™ Medium
CD CHO AGT™ Medium
£108.00 - £5370.00
Specifications
Cell Line | CHO |
---|---|
Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
Culture Type | Suspension Cell Culture |
Form | Powder (AGT) |
Product Type | CD CHO Medium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10003052
|
Gibco™
12490017 |
1 L |
£108.00
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11500436
|
Gibco™
12490025 |
10 L |
£1072.00
10 L |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10389204
|
Gibco™
12490001 |
100 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CD CHO AGT™ (Advanced Granulation Technology) medium is a protein-free, chemically-defined medium optimized for the growth of Chinese hamster ovary (CHO) cells and expression of recombinant proteins in suspension culture. The AGT™ dry media format is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. CD CHO AGT™ medium contains no proteins or peptide components of animal, plant, or synthetic origin, as well as no undefined lysates or hydrolysates.When to Use CD CHO Medium
When to Consider Another Gibco™ Medium
Consider CD FortiCHO™ medium when:
Consider CD OptiCHO™ medium when:
Consider CD DG44 medium when:
Liquid Media Formulations
Gibco™ AGT™ media are also available in a liquid format that offers the same performance and formulation as the AGT™ media format. Gibco™ liquid media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their AGT™ media counterparts.
Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
Specifications
CHO | |
Suspension Cell Culture | |
CD CHO Medium | |
No antibiotics |
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Powder (AGT) | |
Serum-free | |
AGT™, Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.