missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CD FortiCHO™ Medium
CD FortiCHO™ Medium
Brand: Gibco™ A1148301
129.69 GBP valid until 2024-12-31
Use promo code "21885" to get your promotional price.
Code : 50
Additional Details : Weight : 0.05000kg
Description
CD FortiCHO™ medium is specifically designed to offer the highest batch and fed-batch culture performance and yield with recombinant CHO cells in a chemically defined environment. The protein-free, animal origin component-free formulation provides consistency and helps reduce the need to screen for adventitious agents. With CD FortiCHO™ medium cell cultures last 2+ days longer than competitive alternatives, which means decreased HCP (Host Cell Protein) concentration for removal in downstream purification.CD FortiCHO™ medium is fortified to support maximal recombinant CHO cell growth and protein expression in suspension batch culture, offering 120% greater growth and production than select competitive media with simple glucose supplementation. CD FortiCHO™ medium can provide up to 75% greater fed-batch growth and production over glucose-only fed-batch culture for selected cell lines and applications when used with CD EfficientFeed™ C nutrient supplement.
When to Use CD FortiCHO™ Medium
When to Consider Another Gibco™ Medium
Consider CD OptiCHO™ medium when:
Consider CD CHO medium when:
Consider CD DG44 when:
Dry Media Formulations
For early access to CD FortiCHO™ AGT™ medium, please contact Gibco™ Technical Services.
Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
Specifications
Storage: Store in the dark at +2 to +8°C. Guaranteed stable for 12 months when properly stored. |
|
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Suspension Cell Culture | |
CD OptiCHO AGT Medium | |
No Glutamine, No Phenol Red | |
Bottle | |
No antibiotics | |
1 L |
CHO | |
1 X | |
Liquid | |
Sodium Pyruvate, Sodium Bicarbonate | |
Low | |
Sustainable packaging | |
Gibco™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.