missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CD OptiCHO™ AGT™ Medium
CD OptiCHO™ AGT™ Medium
Brand: Gibco™ A1122204
94.75 GBP valid until 2024-12-31
Use promo code "21885" to get your promotional price.
Code : 50
Additional Details : Weight : 0.50000kg
Description
CD OptiCHO™ medium is specifically designed to offer high performance and yield with recombinant CHO cells in a chemically defined fed-batch environment. The protein-free, animal origin free formulation is designed to provide consistency and reduces the need to screen for adventitious agents. CD OptiCHO™ medium builds on Gibco™ CD CHO’s legacy of consistent cell growth and titers, while offering up to 40% better titer and cell growth over select competitors. Thanks to its low osmolality and advanced CHO CD EfficientFeed™ Liquid Nutrient Supplement Kit feeding options, CD OptiCHO™ medium is capable of gaining 2-5 fold increases in fed-batch productivity over batch culture.When to Use CD OptiCHO™ Medium
When to Consider Another Gibco™ Medium
Consider CD FortiCHO™ medium when:
Consider CD CHO medium when:
Consider CD DG44 medium when:
Liquid Media Formulations
Gibco™ AGT™ media are also available in a liquid format that offers the same performance and formulation as the AGT™ media format. Gibco™ liquid media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their AGT™ media counterparts.
Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
Specifications
1 x 1 L, Storage 2-8°C Dark and Dry | |
CHO | |
Suspension Cell Culture | |
CD OptiCHO AGT Medium | |
Non-sterile | |
No Glutamine, No Phenol Red | |
Securitainer Capped Vial | |
Magnesium, Sodium Phosphate | |
Certified | |
1 L | |
Wet Ice |
CO2 | |
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Powder (AGT) | |
Serum-free | |
High Glucose, Sodium Pyruvate, Sodium Bicarbonate | |
Low | |
No antibiotics | |
AGT™, Gibco™ | |
1 L | |
Medium does not support wild type DG44 cells |
For Research Use Only. Not for use in diagnostic procedures.