Learn More
CD140b (PDGFRB) Monoclonal Antibody (APB5), Super Bright™ 600, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen 63-1402-80
Description
Rat Monoclonal Antibody
CD140b (PDGFRB) Monoclonal antibody specifically detects CD140b (PDGFRB) in Mouse samples. It is validated for Flow Cytometry
Specifications
CD140b (PDGFRB) | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P05622 | |
Pdgfrb | |
Affinity chromatography | |
RUO | |
18596 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
APB5 | |
Super Bright 600 | |
Pdgfrb | |
AI528809; Beta platelet-derived growth factor receptor; Beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140b; CD140b antigen; IBGC4; IMF1; JTK12; KOGS; PDGF Receptor beta; Pdgfr; Pdgfr1; PDGFR-1; Pdgfrb; PDGF-R-beta; PENTT; Platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.