missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD147 Monoclonal Antibody (OX114), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA170088
This item is not returnable.
View return policy
Description
MA1-70088 detects CD147 in mouse samples. MA1-70088 has been successfully used in flow cytometry, Western blot and immunohistochemistry procedures. The MA1-70088 immunogen is recombinant md0-2rCD4d3+4.
CD147 (basigin, neurothelin, OX-47, 5A11, CE9, M6, EMMPRIN, extracellular matrix metalloproteinase inducer, TCSF, tumour cell-derived collagenase-stimulatory factor) is a 50-60 kDa glycoprotein with multiple glycosylated forms. CD147 is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is also a member of the immunoglobulin superfamily. The highest level of CD147 expression is on metabolically active cells, such as lymphoblasts, inflammatory cells, brown adipocytes and malignant tumor cells. CD147 has multiple functions, including facilitating of cell surface expression of monocarboxylate transporter proteins and extracellular matrix metalloproteinases, regulation of integrin functions, it plays roles in cell development and activation, fetal development or retinal function.Specifications
CD147 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P18572 | |
BSG | |
Recombinant md0-2rCD4d3+4. | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Western Blot | |
OX114 | |
Unconjugated | |
BSG | |
5A11; 5A11 antigen; 5A11/Basigin; 5A11/Basigin-2; 5F7; AI115436; AI325119; Basic immunoglobulin superfamily; basigin; basigin (Ok blood group); Basigin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); basignin; Basignin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); blood-brain barrier HT7 antigen; BSG; CD147; CD147 antigen; CE9; Collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; glycoprotein CE9; gp 42; gp42/basigin/OX-47/HT7; HT7; HT-7; HT7 antigen; immunoglobulin gene superfamily member; leukocyte activation antigen M6; Membrane glycoprotein gp42; neurothelin; OK; OK blood group antigen; OX-47 antigen; Ox47R; RPE7; TCSF; tumor cell-derived collagenase stimulatory factor; Unknown (protein for MGC:128392); unnamed protein product; UNQ6505/PRO21383 | |
Rat | |
Protein G | |
RUO | |
12215 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |