missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD15 Monoclonal Antibody (MMA (LeuM1)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511789
This item is not returnable.
View return policy
Description
CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determinant (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin's disease.
Specifications
CD15 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P22083 | |
FUT4 | |
U937 histiocytic cell line | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
MMA (LeuM1) | |
Unconjugated | |
FUT4 | |
FUTIV, Lewis X, SSEA-1 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
2526 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD15 Monoclonal Antibody (MMA (LeuM1)), Invitrogen™ > 500 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction