missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16 Monoclonal Antibody (3G8), Alexa Fluor™ 700, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1629
Code : 50
Additional Details : Weight : 0.50000kg
Description
The Alexa Fluor® 700 dye conjugate provides an excellent spectral match to the common long-wavelength excitation sources, with a high extinction coefficient.
CD16 Monoclonal antibody specifically detects CD16 in Human samples. It is validated for Flow Cytometry.Specifications
CD16 | |
Monoclonal | |
Alexa Fluor 700 | |
FCGR3A | |
4833442P21Rik; CD16; CD16-2; CD16A; CD16a antigen; CD16b; CD32; CD32 receptor 2; CDw32; cytolytic trigger molecule G7; Fc fragment of IgG intermediate affinity IV receptor; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG receptor IIIa; Fc fragment of IgG receptor IIIb; Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor; Fc fragment of IgG, low affinity IIIa, receptor (CD16a); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor III; Fc gamma receptor IIIa; Fc gamma receptor III-A; Fc gamma receptor IIIb; Fc gamma RIIIa; Fc gammaRIV; Fc receptor, IgG, low affinity III; Fc receptor, IgG, low affinity IV; Fc receptor-like 3; Fcg receptor III; FCG2; FCG3; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD 16); | |
Mouse | |
purified | |
ASR, ASR/IVD | |
2214, 2215 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
3G8 | |
PBS with BSA and 0.1% sodium azide | |
O75015, P08637 | |
FCGR3A, FCGR3B | |
Human CD16 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |