Learn More
CD16 Monoclonal Antibody (G7), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180267
Code : LD
Additional Details : Weight : 0.01000kg
Description
Clone G7 has been reported to block PBL-mediated antibody-dependent cell cytotoxicity (ADCC) and partially inhibit PMN-mediated ADCC. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Pig CD16, clone G7 recognizes porcine CD16 also known as Fc-gamma RIII or the low affinity IgG (Fc) receptor III.
CD16 encodes a receptor that recognizes the Fc portion of immunoglobulin G and is involved in the clearance of immune complexes from the circulation, as well as other functions such as cellular mediated cytotoxicity and enhancement of virus infections. This gene, FCGR3A, shares a high degree of similarity with another nearby gene, FCGR3B, located on chromosome 1. The receptor encoded by this gene is expressed on natural killer (NK) cells as an integral membrane glycoprotein anchored through a transmembrane peptide, while FCGR3B is expressed on polymorphonuclear neutrophils (PMN) where the receptor is anchored through a phosphatidylinositol (PI) linkage. Mutations in this gene have been associated with immunodeficiency 20 and have been linked to susceptibility to recurrent viral infections, susceptibility to systemic lupus erythematosus, and alloimmune neonatal neutropenia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. Diseases associated with FCGR3A include Immunodeficiency 20 and Herpes Zoster.Specifications
CD16 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
0 | |
FCGR3A | |
Porcine peripheral blood leucocytes. | |
100 Tests | |
Primary | |
Porcine | |
Antibody | |
IgG1 |
Flow Cytometry | |
G7 | |
PE | |
FCGR3A | |
4833442P21Rik; CD16; CD16-2; CD16A; CD16a antigen; CD16b; CD32; CD32 receptor 2; CDw32; cytolytic trigger molecule G7; Fc fragment of IgG intermediate affinity IV receptor; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG receptor IIIa; Fc fragment of IgG receptor IIIb; Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor; Fc fragment of IgG, low affinity IIIa, receptor (CD16a); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor III; Fc gamma receptor IIIa; Fc gamma receptor III-A; Fc gamma receptor IIIb; Fc gamma RIIIa; Fc gammaRIV; Fc receptor, IgG, low affinity III; Fc receptor, IgG, low affinity IV; Fc receptor-like 3; Fcg receptor III; FCG2; FCG3; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD 16); | |
Mouse | |
Protein A | |
RUO | |
397684, 406193 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |