missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD166 Polyclonal Antibody, FITC, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA546875
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) ALCAM is observed, and less than 10% cross-reactivity with rhBCAM, recombinant mouse (rm) OCAM, and rmMAdCAM-1 is observed.
CD166 Polyclonal antibody specifically detects CD166 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD166 | |
Polyclonal | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q61490 | |
ALCAM | |
Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166 Trp28-Lys527 | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
FITC | |
ALCAM | |
activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
Goat | |
Antigen affinity chromatography | |
RUO | |
11658 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |