missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD177 Monoclonal Antibody (MEM-166), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15774
This item is not returnable.
View return policy
Description
CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregcy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).Specifications
CD177 | |
Monoclonal | |
PE | |
CD177 | |
HNA-2a, HNA2A, NB1, NB1 GP, PRV-1, PRV1, CD177 antigen, NB1 glycoprotein, cell surface receptor, human neutrophil alloantigen 2a, polycythemia rubra vera 1, polycythemia rubra vera protein 1 | |
Mouse | |
Purified | |
RUO | |
57126 | |
4° C | |
Liquid |
Flow Cytometry | |
MEM-166 | |
PBS with 0.2% BSA and 15mM sodium azide | |
Q8N6Q3 | |
CD177 | |
Human CD177 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |