Learn More
CD18 Monoclonal Antibody (10E12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA515517
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15517 targets CD18 in indirect ELISA, IF applications and shows reactivity with Human and mouse samples. The MA5-15517 immunogen is purified recombinant fragment of CD18 expressed in E. Coli. MA5-15517 detects CD18 which has a predicted molecular weight of approximately 85kDa.
CD18 integrin beta 2 subunit is a 90 kDa type I transmembrane protein expressed on all leucocytes. CD18 can combine with integrin molecules CD11a-c to form heterodimers at the cell surface, and these heterodimers are known to participate in the process of cell adhesion as well as cell-surface mediated signaling. CD18 forms heterodimers with four types of CD11 molecule to constitute leukocyte (beta2) integrins: alphaLbeta2 (CD11a/CD18, LFA-1), alphaMbeta2 (CD11b/CD18, Mac-1, CR3), alphaXbeta2 (CD11c/CD18) and alphaDbeta2 (CD11d/CD18). In most cases, the response mediated by the integrin is a composite of the functions of its individual subunits, and these integrins are essential for proper leukocyte migration, mediating intercellular contacts. Absence of CD18 leads to leukocyte adhesion deficiency-1, severe reduction of CD18 expression leads to the development of a psoriasiform skin disease. CD18 is also a target of Mannheimia (Pasteurella) haemolytica leukotoxin and is sufficient to mediate leukotoxin-mediated cytolysis. Defects in the CD18 gene are the cause of leukocyte adhesion deficiency type I (LAD1). Two transcript variants encoding the CD18 protein have been identified.Specifications
CD18 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P05107, P11835 | |
ITGB2 | |
Purified recombinant fragment of CD18 expressed in E. Coli. | |
RUO | |
16414, 3689 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry | |
10E12 | |
Unconjugated | |
ITGB2 | |
2E6; AI528527; antigen CD18; B-2 integrin; CD18; CD18 leukocyte adhesion molecule; CD18 subunit; cell surface adhesion glycoprotein (LFA-1/CR3/P150,959 beta subunit precursor); cell surface adhesion glycoproteins LFA-1/CR3/p150,95 subunit beta; complement component 3 receptor 3 and 4 subunit; complement receptor C3 beta-subunit; complement receptor C3 subunit beta; integrin beta 2; integrin beta chain, beta 2; integrin beta-2; integrin subunit beta 2; integrin, beta 2; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; macrophage antigen 1 (mac-1) beta subunit); integrin, beta 2 (antigen CD18 subunit (p95), lymphocyte function-associated antigen 1, integrin B2); integrin, beta 2 (complement component 3 receptor 3 and 4 subunit); ITGB2; LAD; LCAMB; Leu-CAM receptor; leukocyte cell adhesion molecule CD18; leukocyte surface protein; leukocyte-associated antigens CD18/11A, CD18/11B, | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |