Learn More
CD19 Monoclonal Antibody (SJ25-C1), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1905
Code : 50
Additional Details : Weight : 0.50000kg
Description
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Monoclonal | |
APC | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Mouse | |
purified | |
ASR | |
930 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
SJ25-C1 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P15391 | |
Cd19 | |
Nalm 1 Human cell line | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |